Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
Brand:  Thermo Scientific™ SPD300DDAA-230
 View more versions of this product
Product Code. 15946181
  • £35598.87 / Each
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
  •  For strong acids and bases used in combinatorial chemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UL Listed to US and Canadian requirements. CE marked
SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor
230 V 50 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits >