Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Large Capacity Concentrators
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Brand: Thermo Scientific™ MPTR8-210
This item is not returnable.
View return policy
Description
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
Specifications
Microwell Plates: 8 Deep-well, Large capacity models | |
SPD210, SPD2030, SPD300DDA (requires RAD200) | |
MPTR8-210 |
Rotor | |
8 Microwell Plates | |
Traditional |