missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Rotors for Large Capacity Concentrators

SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.

Brand:  Thermo Scientific™ RH60-17-100

 View more versions of this product

Product Code. 11788559

  • £1322.25 / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.

Circular Rotors:

  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Centrifuge Tubes: 30 x 15L conical, Large capacity models
SPD210, SPD2030, SPD300DDA (requires RAD200)
RH60-17-100
15 mL
Rotor
30 15 mL Conical Centrifuge Tubes
Traditional
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers