Learn More
Scandium(III) oxide, REacton™, 99.998% (metals basis), REM 50ppm max, Thermo Scientific Chemicals
Description
A high melting white solid used in high-temperature systems (for its resistance to heat and thermal shock), electronic ceramics, and glass composition. Suitable for vacuum deposition applications. Also used to make scandium fluoride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2485°C |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.998% (metals basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 137.91 |
Odor | Odorless |
Refractive Index | 1.964 |
Chemical Name or Material | Scandium(III) oxide, REM 50ppm max. |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
L21Y073 | Certificate of Analysis | 13/01/2025 | 010916.03,010916.06 |
Z03H034 | Certificate of Analysis | 12/04/2022 | 010916.03,010916.06 |
L27S027 | Certificate of Analysis | 07/12/2021 | 010916.03,010916.06 |
N29C045 | Certificate of Analysis | 07/12/2021 | 010916.03,010916.06 |
B16Y029 | Certificate of Analysis | 07/12/2021 | 010916.03,010916.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.