Learn More
Scandium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™
£76.58 - £277.00
Chemical Identifiers
CAS | 14066-05-8 |
---|---|
Molecular Formula | Cl3O12Sc |
Molecular Weight (g/mol) | 343.294 |
MDL Number | MFCD00243289 |
InChI Key | UBVKRMCXDCYZQK-UHFFFAOYSA-K |
Synonym | scandium perchlorate, scandium 3+ perchlorate, acmc-20ajr3, scandium 3+ ion triperchlorate, trihyperchloric acid scandium salt, scandium iii perchlorate solution, scandium 3+ triperchlorateShow More |
PubChem CID | 11013318 |
IUPAC Name | scandium(3+);triperchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Sc+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430311
|
Thermo Scientific Alfa Aesar
040579.NA |
2 g |
£76.58
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440311
|
Thermo Scientific Alfa Aesar
040579.NC |
10 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
14066-05-8 | |
343.294 | |
UBVKRMCXDCYZQK-UHFFFAOYSA-K | |
11013318 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Sc+3] |
Cl3O12Sc | |
MFCD00243289 | |
scandium perchlorate, scandium 3+ perchlorate, acmc-20ajr3, scandium 3+ ion triperchlorate, trihyperchloric acid scandium salt, scandium iii perchlorate solution, scandium 3+ triperchlorateShow More | |
scandium(3+);triperchlorate |
Specifications
14066-05-8 | |
Cl3O12Sc | |
UN3211 | |
Insoluble in water. | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Sc+3] | |
343.294 | |
451.42 (343.30 Anhydrous) | |
50% w/w aq. soln. | |
Scandium(III) perchlorate |
Liquid | |
MFCD00243289 | |
scandium perchlorate, scandium 3+ perchlorate, acmc-20ajr3, scandium 3+ ion triperchlorate, trihyperchloric acid scandium salt, scandium iii perchlorate solution, scandium 3+ triperchlorate | |
UBVKRMCXDCYZQK-UHFFFAOYSA-K | |
scandium(3+);triperchlorate | |
11013318 | |
Reagent | |
Odorless |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
missing translation for 'einecsNumber' : 237-914-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.