Learn More
Eppendorf™ Scientific Shaker Universal Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand:  Eppendorf™ M1250-9920
 View more versions of this product
Product Code. 11355994
  • £1345.36 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Requires : Clamps (sold separately)
Description
Description
  • Accept a variety of accessory clamps, test tube racks and gel trays for greater flexibility
  • Hold combinations of flasks or test tube racks
  • With pre-drilled holes
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal Platform
18 x 30 in. (457 x 762 mm)
30 in.
76 cm
18 in.
46 cm
Pre-drilled holes
Aluminum
18 in.
457 mm
30 in.
762 mm
Innova 43/43R, 2300, series 4300 and 4400, E10, E25, E25R, C10, C25/C25KC shaker
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Scientific Shaker Universal Platforms > For Innova 43/43R, 2300, Series 4300 and 4400, E10, E25, E25R, C10, C25/C25KC shakers