Learn More
Eppendorf™ Scientific Shaker Universal Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
£537.14 - £1400.58
Specifications
Product Type | Universal Platform |
---|---|
Includes | Pre-drilled holes |
Requires: Clamps (sold separately)
Product Code | Brand | Length (English) Platform | Width (English) Platform | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) Platform | Width (English) Platform | For Use With (Equipment) | Price | Quantity & Availability | ||||
11692764
|
Eppendorf™
PTL-386 |
24 in. | 36 in. | Innova 2350 shaker |
£1400.58
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
||||
11365964
|
Eppendorf™
M1250-9902 |
18 in. | 18 in. | For Innova, I-Series, Excella shaker |
£978.90
Each |
In Stock
Log in to see stock. |
||||
11355994
|
Eppendorf™
M1250-9920 |
30 in. | 18 in. | Innova 43/43R, 2300, series 4300 and 4400, E10, E25, E25R, C10, C25/C25KC shaker |
£1345.36
Each |
In Stock
Log in to see stock. |
||||
11319844
|
Eppendorf™
M1190-9900 |
16 in. | 12 in. | Innova Portable Model 2050 |
£657.62
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||
12596666
|
Eppendorf™
M1324-9904 |
30 in. | 18 in. | Innova I26/I-26R shaker |
£1325.28
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11309844
|
Eppendorf™
M1001-0240 |
13 in. | 11 in. | Innova 2000, Excella E1/E2 |
£537.14
Each |
In Stock
Log in to see stock. |
||||
11309834
|
Eppendorf™
M1194-9912 |
24 in. | 18 in. | Innova Model 2150 shaker |
£1044.16
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Accept a variety of accessory clamps, test tube racks and gel trays for greater flexibility
- Hold combinations of flasks or test tube racks
- With pre-drilled holes
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Universal Platform | |
Pre-drilled holes |