missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23024-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregcy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.Specifications
SCP3 | |
Polyclonal | |
Unconjugated | |
SYCP3 | |
COR1, SCP 3, SCP3, SYCP3, synaptonemal complex protein 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20962, 25561, 50511 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70281, Q63520, Q8IZU3 | |
Sycp3 | |
SCP3 Fusion Protein Ag19330 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |