missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | SCP3 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16842165
|
Proteintech
23024-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16832165
|
Proteintech
23024-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregcy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.Specifications
SCP3 | |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P70281, Q63520, Q8IZU3 | |
20962, 25561, 50511 | |
SCP3 Fusion Protein Ag19330 | |
Primary | |
-20°C | |
SYCP3 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COR1, SCP 3, SCP3, SYCP3, synaptonemal complex protein 3 | |
Sycp3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |