missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Screw Neck Vial, 15-425 thread, Amber Glass

Screw Neck Vials for storage purposes made out of 1st hydrolytic class glass.

Brand:  Fisherbrand™ 15 09 1774

26.76 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 11596064

  • £37.85 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Screw Neck
  • 15-425
  • Flat bottom
  • Glass
  • Amber
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amber
Glass
Storage
CleanPack, Polypropylene Box
8 mL
61 mm
ND15
16.6 mm
8.9 mL
Screw
800 μL
Flat Bottom
100
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.