missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Screw-Top Bottle, Square, Clear, Soda-Lime Glass, Narrow Mouth, High form DIN thread
Square clear soda lime glass bottles, narrow mouth.
£46.80 - £113.00
Spécification
For Use With (Application) | Storage of samples and reagents; Sample collection; Transport and shipping |
---|---|
Material | Soda Lime Glass |
Quantity | 10/Pk. |
Autoclavable | Not Autoclavable |
Code produit | Marque | Capacity (Metric) | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Capacity (Metric) | Prix | Quantité et disponibilité | |||||
12850924
|
DWK Life Sciences
238102456 |
100 mL |
£46.80
Pack of 10 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
11776918
|
DWK Life Sciences
238103658 |
250 mL |
£81.05
Pack of 10 |
Estimated Shipment: 31-10-2024
Log in to see stock. |
|||||
11706928
|
DWK Life Sciences
238104457 |
500 mL | |||||||
11716928
|
DWK Life Sciences
238105453 |
1000 mL | |||||||
Description
Square clear soda lime (Type 3) glass bottles, narrow mouth, high form DIN thread.
- Square shape for efficient packing
Spécification
Storage of samples and reagents; Sample collection; Transport and shipping | |
Soda Lime Glass | |
10/Pk. | |
Not Autoclavable |