Learn More
GeneBLAzer™ SCTR-CRE-bla CHO-K1 Cells
GeneBLAzer™ SCTR-CRE-bla CHO-K1 Cells contain the human Secretin Receptor (SCTR).
Brand: GeneBLAzer™ K1805
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: SCTR
- Accession Number: NM_002980.2
- Population Status: Pool of Stable Cells
- GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells contain the human Secretin Receptor (SCTR) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. no. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.
- The GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells are functionally validated for Z' and EC50concentrations of Secretin. In addition, GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Liquid Nitrogen | |
GPCRs | |
8 x 106 Cells |
Fluorescent | |
SCTR-CRE-bla CHO-K1 cells, SCTR-CRE-bla CHO-K1, Certificate of Analysis | |
Dividing Cells |