Learn More
Invitrogen™ SDF-1 alpha/CXCL12 Mouse ELISA Kit
Sandwich ELISA Kit
£506.00 - £4370.00
Specifications
Accession Number | P40224 |
---|---|
Assay Sensitivity | 3 pg/mL |
Description | SDF-1 alpha CXCL12 ELISA Kit, 96 Assays, 1 Kit, Mouse Species, Streptavidin-HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of SDF-1 alpha CXCL12 in Samples Using a Microplate Rea |
Detectable Analytes | SDF-1 alpha CXCL12, mouse SDF-1 |
Assay | Solid-phase sandwich ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15445253
|
Invitrogen™
EMCXCL12 |
96 Tests |
£506.00
1 set |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17165709
|
Invitrogen™
EMCXCL12X5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
18507924
|
Invitrogen™
EMCXCL12X10 |
10 x 96 Tests |
£4370.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Mouse SDF-1 alpha/CXCL12 ELISA quantitates Ms SDF-1α in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms SDF-1α. Principle of the method The Mouse SDF-1α solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
P40224 | |
SDF-1 alpha CXCL12 ELISA Kit, 96 Assays, 1 Kit, Mouse Species, Streptavidin-HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of SDF-1 alpha CXCL12 in Samples Using a Microplate Rea | |
Solid-phase sandwich ELISA | |
Colorimetric | |
Chemokine & Receptors | |
Mouse | |
Colorimetric Microplate Reader | |
Pbsf,Scyb12,Sdf1,Tlsf,Tpar1 | |
<12% | |
HRP | |
Immunology | |
Pbsf, Scyb12, Sdf1, Tlsf, Tpar1 | |
1 hr. 20 min. |
3 pg/mL | |
SDF-1 alpha CXCL12, mouse SDF-1 | |
Biotin | |
ELISA Kit | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin, Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3 | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
20315 | |
Mouse | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.