Learn More
Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Sebacoyl chloride, 92%, C10H16Cl2O2, CAS Number-111-19-3, decanedioic acid chloride, sebacoyl chloride, cp, octane-1,8-dicarbonyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, sebacoyl chloride, sebacoylchloride, acmc-1byrk, decanedioyl chloride, 500g, 90% min. (GC)
£50.79 - £251.90
Chemical Identifiers
CAS | 111-19-3 |
---|---|
Molecular Formula | C10H16Cl2O2 |
Molecular Weight (g/mol) | 239.14 |
MDL Number | MFCD00000770 |
InChI Key | WMPOZLHMGVKUEJ-UHFFFAOYSA-N |
Synonym | sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chlorideShow More |
PubChem CID | 66072 |
IUPAC Name | decanedioyl dichloride |
SMILES | C(CCCCC(=O)Cl)CCCC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10668313
|
Thermo Scientific Acros
294900500 |
50 g | Glass bottle |
£50.79
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10422261
|
Thermo Scientific Acros
294902500 |
250 g | Glass bottle |
£150.75
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15790849
|
Thermo Scientific Acros
294905000 |
500 g | Glass bottle |
£251.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-19-3 | |
239.14 | |
WMPOZLHMGVKUEJ-UHFFFAOYSA-N | |
66072 | |
C(CCCCC(=O)Cl)CCCC(=O)Cl |
C10H16Cl2O2 | |
MFCD00000770 | |
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chlorideShow More | |
decanedioyl dichloride |
Specifications
111-19-3 | |
White to Beige | |
220.0°C (75.0 mmHg) | |
Authentic | |
C10H16Cl2O2 | |
1.4670 to 1.4700 | |
02, 719 | |
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride | |
WMPOZLHMGVKUEJ-UHFFFAOYSA-N | |
decanedioyl dichloride | |
66072 | |
92% | |
Liquid |
-2.5°C | |
1.1210g/mL | |
>110°C | |
90% min. (GC) | |
ClCO(CH2)8COCl | |
MFCD00000770 | |
1.121 | |
Solubility in water: may decompose. Other solubilities: soluble in hydrocarbons and ethers | |
C(CCCCC(=O)Cl)CCCC(=O)Cl | |
239.14 | |
239.14 | |
Technical | |
Sebacoyl chloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-843-4
missing translation for 'rtecsNumber' : HD8454250
missing translation for 'tsca' : TSCA
Recommended Storage : Product may develop some turbidity or precipitate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.