missing translation for 'onlineSavingsMsg'
Learn More

sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals

£22.71 - £108.54

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10448630
View Documents Promotion Details
Thermo Scientific Acros
107700010
1 L Glass Bottle
£22.71
1 L
Please to purchase this item. Need a web account? Register with us today!
10765361
View Documents Promotion Details
Thermo Scientific Acros
107700025
2.5 L Glass Bottle
£38.79
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10512744
View Documents Promotion Details
Thermo Scientific Acros
107700100
10 L Plastic Drum
£108.54
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78-92-2
74.12
BTANRVKWQNVYAZ-UHFFFAOYNA-N
6568
butan-2-ol
C4H10O
MFCD00004569
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
CHEBI:35687
CCC(C)O

Specifications

78-92-2
Conforms to Ref Report
1.3960 to 1.3980 (20°C, 589nm)
0.005% max.
C2H5CH(OH)CH3
01, 371
15, 1543
BTANRVKWQNVYAZ-UHFFFAOYNA-N
butan-2-ol
6568
74.12
Extra Pure
0.8100g/mL
Liquid
99%
C4H10O
MFCD00004569
0.5% max.
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
CCC(C)O
74.12
CHEBI:35687
≥99%
sec-Butanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • sec-Butanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only