missing translation for 'onlineSavingsMsg'
Learn More
Selenium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11370759

Selenium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11370759
5 g
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11370759

Brand: Thermo Scientific Alfa Aesar 010601.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-49-2
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]

Specifications

Physical Form Powder
Quantity 5 g
Assay Percent Range 99.5% (Metals basis)
Odor Odorless
Appearance Various colors powder
Density 4.28g/cm3
Chemical Name or Material Selenium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN02K054 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number010601.06,010601.18,010601.30
Lot NumberT30F050 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog Number010601.06,010601.18,010601.30
Lot NumberU31I008 Certificate TypeCertificate of Analysis Date21/09/2022 Catalog Number010598.09,010601.06,010598.18,010598.90
Lot NumberZ14H032 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010601.06,010601.18,010601.30
Lot NumberZ06H051 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010598.09,010601.06,010598.18,010598.90

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Selenium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.