Learn More
Brady™ Self-laminating, Self-extinguishing Tedlar™ Wrap Around Wire and Cable Labels for M6/M7 Printers
The translucent polyvinylflouride material with permanent adhesive of these labels has high flexibility for wrapping and small diameter wires.
Brand:  Brady™ 173339
Additional Details : Weight : 0.11100kg
 View more versions of this product
Product Code. 17839224
  • £122.00 / Pack of 250
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • Self-extinguishing material is ideal for wire and cable marking applications in the aerospace, defense and mass transit industries
  • High solvent resistance
  • Self-laminating
  • RoHS compatibility Pass 2011/65/EU
  • Made of B-642 polyvinylfluoride
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Clear, White
Die-cut
Single-side Printable
Labels
Data and Telecommunications Labeling, Wire and Cable Labeling
No
Rectangle
-70°C to +120°C
0.055 mm
1.5 in.
25.4 mm
25.4 mm
38.1 mm
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155
None
Matte
PVF
Dot Matrix, Thermal Transfer
Non-sterile
M710, BMP71
250 Labels
-94°F to +248°F
0.0022 in.
1.5 x 1 in.
1 in.
1 in.
0.5 in.
12.7 mm