Learn More
Selpercatinib, MedChemExpress
MedChemExpress Selpercatinib (LOXO-292) is a potent, selective RET kinase inhibitor with IC50 values of 14.0 nM, 24.1 nM, and 530.7 nM for RET (WT), RET (V804M), and RET (G810R), respectively. Selpercatinib has anticancer activity.
Brand:  Medchem Express HY-114370
Encompass_Preferred
 View more versions of this product
Product Code. 16455616
  • £744.15 / 100mg
Explore more special offers
Description
Description
Selpercatinib (LOXO-292) is a potent, selective RET kinase inhibitor with IC50 values of 14.0 nM, 24.1 nM, and 530.7 nM for RET (WT), RET (V804M), and RET (G810R), respectively. Selpercatinib has anticancer activity.
Specifications
Specifications
Selpercatinib
White
C29H31N7O3
LOXO-292
N#CC1=C2C(C3=CC=C(N4CC(C5)N(CC6=CC=C(OC)N=C6)C5C4)N=C3)=CC(OCC(C)(O)C)=CN2N=C1
525.6
Research
Cancer-Kinase/protease
2152628-33-4
Research
100 mg
DMSO : 62.5 mg/mL (118.91 mM; Need ultrasonic)
525.6
98.06%
Solid