Learn More
Selpercatinib, MedChemExpress
MedChemExpress Selpercatinib (LOXO-292) is a potent, selective RET kinase inhibitor with IC50 values of 14.0 nM, 24.1 nM, and 530.7 nM for RET (WT), RET (V804M), and RET (G810R), respectively. Selpercatinib has anticancer activity.
Brand: Medchem Express HY-114370
This item is not returnable.
View return policy
Description
Selpercatinib (LOXO-292) is a potent, selective RET kinase inhibitor with IC50 values of 14.0 nM, 24.1 nM, and 530.7 nM for RET (WT), RET (V804M), and RET (G810R), respectively. Selpercatinib has anticancer activity.Specifications
Selpercatinib | |
White | |
C29H31N7O3 | |
LOXO-292 | |
N#CC1=C2C(C3=CC=C(N4CC(C5)N(CC6=CC=C(OC)N=C6)C5C4)N=C3)=CC(OCC(C)(O)C)=CN2N=C1 | |
525.6 | |
Research | |
Cancer-Kinase/protease |
2152628-33-4 | |
Research | |
5 mg | |
DMSO : 62.5 mg/mL (118.91 mM; Need ultrasonic) | |
525.6 | |
98.06% | |
Solid |