Learn More
Semi-Volatile Calibration Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep BIG-BN-2-5
Code : 04
Additional Details : Weight : 0.10000kg
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,2,4-Trichlorobenzene | 120-82-1 | 2000μg/mL |
1,2-Dichlorobenzene | 95-50-1 | 2000μg/mL |
1,3-Dichlorobenzene | 541-73-1 | 2000μg/mL |
1,4-Dichlorobenzene | 106-46-7 | 2000μg/mL |
2,4-Dinitrotoluene | 121-14-2 | 2000μg/mL |
2,6-Dinitrotoluene | 606-20-2 | 2000μg/mL |
2-Chloronaphthalene | 91-58-7 | 2000μg/mL |
2-Methylnaphthalene | 91-57-6 | 2000μg/mL |
2-Nitroaniline | 88-74-4 | 2000μg/mL |
3-Nitroaniline | 99-09-2 | 2000μg/mL |
4-Bromodiphenyl ether | 101-55-3 | 2000μg/mL |
4-Chloroaniline (p-Chloroaniline) | 106-47-8 | 2000μg/mL |
4-Chlorophenyl-phenyl ether | 7005-72-3 | 2000μg/mL |
4-Nitroaniline | 100-01-6 | 2000μg/mL |
Acenaphthene | 83-32-9 | 2000μg/mL |
Acenaphthylene | 208-96-8 | 2000μg/mL |
Anthracene | 120-12-7 | 2000μg/mL |
Benz(a)anthracene | 56-55-3 | 2000μg/mL |
Benzo(a)pyrene | 50-32-8 | 2000μg/mL |
Benzo(b)fluoranthene | 205-99-2 | 2000μg/mL |
Benzo(g,h,i)perylene | 191-24-2 | 2000μg/mL |
Benzo(k)fluoranthene | 207-08-9 | 2000μg/mL |
Benzyl alcohol | 100-51-6 | 2000μg/mL |
Bis(2-chloro-1-methylethyl) ether | 108-60-1 | 2000μg/mL |
bis(2-Chloroethoxy)methane | 111-91-1 | 2000μg/mL |
bis(2-Chloroethyl)ether | 111-44-4 | 2000μg/mL |
Bis(2-Ethylhexyl)phthalate | 117-81-7 | 2000μg/mL |
Butylbenzyl phthalate | 85-68-7 | 2000μg/mL |
Chrysene | 218-01-9 | 2000μg/mL |
Dibenz(a,h)anthracene | 53-70-3 | 2000μg/mL |
Dibenzofuran | 132-64-9 | 2000μg/mL |
Diethyl phthalate | 84-66-2 | 2000μg/mL |
Dimethyl phthalate | 131-11-3 | 2000μg/mL |
Di-n-butyl phthalate | 84-74-2 | 2000μg/mL |
Di-n-octyl phthalate | 117-84-0 | 2000μg/mL |
Fluoranthene | 206-44-0 | 2000μg/mL |
Fluorene | 86-73-7 | 2000μg/mL |
Hexachlorobenzene | 118-74-1 | 2000μg/mL |
Hexachlorobutadiene | 87-68-3 | 2000μg/mL |
Hexachlorocyclopentadiene | 77-47-4 | 2000μg/mL |
Hexachloroethane | 67-72-1 | 2000μg/mL |
Indeno(1,2,3-c,d)pyrene | 193-39-5 | 2000μg/mL |
Isophorone | 78-59-1 | 2000μg/mL |
Naphthalene | 91-20-3 | 2000μg/mL |
Nitrobenzene | 98-95-3 | 2000μg/mL |
N-Nitrosodi-n-propylamine | 621-64-7 | 2000μg/mL |
N-Nitrosodiphenylamine | 86-30-6 | 2000μg/mL |
Phenanthrene | 85-01-8 | 2000μg/mL |
Pyrene | 129-00-0 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Semi-volatile Calibration Standard | |
5 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
For US EPA Methods 625, 8270, CLP |
5mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
Components in Methylene Chloride:Benzene at 2000μg/mL Acenaphthene Dimethyl Phthalate Acenaphthylene Di-n-butyl Phthalate Anthracene 2,4-Dinitrotoluene Benzo(a)anthracene 2,6-Dinitrotoluene Benzo(a)pyrene Di-n-octyl Phthalate Benzo(b)fluoranthene Fluoranthene Benzo(g,h,i)perylene Fluorene Benzo(k)fluoranthene Hexachlorobenzene Benzyl Alcohol Hexachlorobutadiene bis(2-chloroethoxy)methane Hexachlorocyclopentadiene bis(2-chloroethyl)ether Hexachloroethane bis(2-chloroisopropyl)ether Indeno(1,2,3-cd)pyrene bis(2-ethylhexyl)phthalate Isophorone 4-Bromophenyl phenyl ether 2-Methylnaphthalene Butylbe |
|
MeCl:Benzene |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1A
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dichloromethane, benzene, 1,4-dichlorobenzene, 2,4-dinitrotoluene, 4-bromodiphenyl ether
Product intended for laboratory use only