missing translation for 'onlineSavingsMsg'
Learn More

Semi-Volatile Calibration Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

£273.00 - £1170.00

Specifications

Chemical Name or Material Semi-volatile Calibration Standard
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Methods For US EPA Methods 625, 8270, CLP
Solvent or Matrix MeCl:Benzene
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12797496
View Documents
SPEX Certiprep
BIG-BN-2
1 mL 2mL amber prescored glass ampule
£273.00
1mL
Please to purchase this item. Need a web account? Register with us today!
12707506
View Documents
SPEX Certiprep
BIG-BN-2-5
5 mL 5mL amber prescored glass ampule This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2,4-Trichlorobenzene120-82-12000μg/mL
1,2-Dichlorobenzene95-50-12000μg/mL
1,3-Dichlorobenzene541-73-12000μg/mL
1,4-Dichlorobenzene106-46-72000μg/mL
2,4-Dinitrotoluene121-14-22000μg/mL
2,6-Dinitrotoluene606-20-22000μg/mL
2-Chloronaphthalene91-58-72000μg/mL
2-Methylnaphthalene91-57-62000μg/mL
2-Nitroaniline88-74-42000μg/mL
3-Nitroaniline99-09-22000μg/mL
4-Bromodiphenyl ether101-55-32000μg/mL
4-Chloroaniline (p-Chloroaniline)106-47-82000μg/mL
4-Chlorophenyl-phenyl ether7005-72-32000μg/mL
4-Nitroaniline100-01-62000μg/mL
Acenaphthene83-32-92000μg/mL
Acenaphthylene208-96-82000μg/mL
Anthracene120-12-72000μg/mL
Benz(a)anthracene56-55-32000μg/mL
Benzo(a)pyrene50-32-82000μg/mL
Benzo(b)fluoranthene205-99-22000μg/mL
Benzo(g,h,i)perylene191-24-22000μg/mL
Benzo(k)fluoranthene207-08-92000μg/mL
Benzyl alcohol100-51-62000μg/mL
Bis(2-chloro-1-methylethyl) ether108-60-12000μg/mL
bis(2-Chloroethoxy)methane111-91-12000μg/mL
bis(2-Chloroethyl)ether111-44-42000μg/mL
Bis(2-Ethylhexyl)phthalate117-81-72000μg/mL
Butylbenzyl phthalate85-68-72000μg/mL
Chrysene218-01-92000μg/mL
Dibenz(a,h)anthracene53-70-32000μg/mL
Dibenzofuran132-64-92000μg/mL
Diethyl phthalate84-66-22000μg/mL
Dimethyl phthalate131-11-32000μg/mL
Di-n-butyl phthalate84-74-22000μg/mL
Di-n-octyl phthalate117-84-02000μg/mL
Fluoranthene206-44-02000μg/mL
Fluorene86-73-72000μg/mL
Hexachlorobenzene118-74-12000μg/mL
Hexachlorobutadiene87-68-32000μg/mL
Hexachlorocyclopentadiene77-47-42000μg/mL
Hexachloroethane67-72-12000μg/mL
Indeno(1,2,3-c,d)pyrene193-39-52000μg/mL
Isophorone78-59-12000μg/mL
Naphthalene91-20-32000μg/mL
Nitrobenzene98-95-32000μg/mL
N-Nitrosodi-n-propylamine621-64-72000μg/mL
N-Nitrosodiphenylamine86-30-62000μg/mL
Phenanthrene85-01-82000μg/mL
Pyrene129-00-02000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Semi-volatile Calibration Standard
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
MeCl:Benzene
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For US EPA Methods 625, 8270, CLP
Videos
Safety and Handling

Safety and Handling

Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.

missing translation for 'dotInformation' : Nonhazardous

Recommended Storage : Ambient

Product Identifier
  • Semi-volatile Calibration Standard

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : anthracene, hexachlorobuta-1,3-diene, 1,2,4-trichlorobenzene

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.