Learn More
Julabo™ SemiChill Recirculating Cooling Baths with Air Compressor
Operational temperature ranges from +5 to +35 °C. Julabo™ SemiChill Recirculating Cooling Baths with Air Compressor can be modified to provide heater capacities up to 5 kW.
Brand:  Julabo™ 950002503P3H0
 View more versions of this product
Product Code. 11914838
  • £20485.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Space-saving, no lateral ventilation
  • Handles and castor make relocation easy
  • Industrial grade mains power switch and emergency cut-off
  • Pressure gauge
  • Liquid level indicator
  • Front filling port
  • Low noise level
  • PID temperature control
  • ATC3 3-point calibration
  • Early warning system for low liquid level
  • Warning and safety functions
  • Modular design allows selection between different options
  • Air cooled model
Specifications
Specifications
21 to 33 L
0.9kW at -10°C, 1.5kW at 0°C, 2.5kW at 20°C (Medium Ethanol)
For industrial applications
1 kW
SC2500a
±0.1°C
49 cm
123 kg
230 V
CE, Classification III (FL) according to DIN 12876-1
33 L/min.
5°C to 40°C
50.8 psi
R404A
62 cm
105 cm
230 V 60 Hz 10 A
60 Hz