Learn More
Cytiva (formerly Pall Lab) Sentino™ Filters Funnel, Gamma Irradiated
Sentino™ Filter Funnels are single-use funnels designed for use with the Sentino™ pump with a press-fit seal, membrane guides and forceps access point allowing for simple set up and membrane removal.
Brand:  Cytiva (formerly Pall Lab) 4871
Additional Details : Weight : 1.18750kg
 View more versions of this product
Product Code. 17134431
  • £152.00 / Pack of 80
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
  • For research use only
Specifications
Specifications
250 mL
20/Sleeve, 80/Pk.
Filtering Funnel
Polypropylene
Sterile