Learn More
Cytiva (formerly Pall Lab) Sentino™ Metricel™ Black Membrane
Filter dispense packs for Sentino dispenser are ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes.
Brand:  Cytiva (formerly Pall Lab) 68125
 View more versions of this product
Product Code. 17194591
  • £149.00 / Pack of 200
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Filter dispensers are ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes
  • Designed to aseptically dispense one disc of membrane at the touch of a button
  • Gamma irradiated and ready to use
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Metricel Black Membrane Modified PES
Non-sterile
47 mm
0.8 μm
1000/Pk., Dispenser Pack of 200