missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephacryl™ S-400 High Resolution Media

Hydrophilic, rigid allyl dextran/bisacrylamide matrix for fast and reproducible purification of proteins, polysaccharides and other macromolecules

Brand:  Cytiva 17-0609-10

Code : GE

Additional Details : Weight : 0.23700kg


Product Code. 11520794

  • £166.00 / 150mL
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

Chromatography media offer good recovery and low nonspecific adsorption. Enable gel filtration at laboratory and industrial scale, and may be purchased in prepacked columns.

  • For separating polysaccharides, macromolecules with extended structures, and small plasmids
  • Excellent purification over a wide molecular weight range
  • High reproducibility due to long-term chemical and physical stability
  • High flow rates and recoveries
  • Well-suited to industrial-scale use
  • Storage conditions: 20% ethanol; 4° to 30°C
  • Available in a variety of selectivities and volumes

Chemical Stability (stable in all common buffers):

  • 1M acetic acid, 8M urea
  • 6M guanidine-HCl, 1% SDS
  • 2M NaCl, 24% ethanol
  • 30% propanol, 30% acetonitrile
  • 0.5M NaOH (for cleaning)
pH Stability
  • 3 to 11 (long-term and working)
  • 2 to 13 (short-term)
Physical Stability:
  • Negligible volume variation due to changes in pH or ionic strength
  • Autoclavability: At 121°C, pH 7 for 30 minutes (gel only)
  • Antimicrobial agent: 20% ethanol

Specifications

Specifications

150mL
25 to 75 μm
Sephacryl HR Media
40 to 140cm
5.9 in.
100kPa
Allyl Dextran and N, N'-methylenebisacrylamide
4° to 30°C
>150 cm/hr.
19 to 230L
Solid Spherical Bead
Allow fast and reproducible purification of proteins, polysaccharides and other macromolecules by gel filtration at laboratory and industrial scale
15cm
3 to 11
Polysaccharides, macromolecules and small plasmids
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sephacryl S-400 High Resolution, 150 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers