missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G-100 Media
Commonly used for desalting and buffer exchange of very large biomolecules
£534.00 - £669.00
Specifications
Quantity | 100g |
---|---|
Bed Volume | 15 to 20mL/g in Distilled Water |
Product Type | Sephadex G-100 Media |
Format | Solid Bead |
For Use With (Application) | Useful for desalting and Buffer exchange of very large biomolecules |
Product Code | Brand | Flow Rate | Particle Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Particle Size | Price | Quantity & Availability | ||||
11524885
|
Cytiva
17-0060-01 |
4.2 mL/min. | 100 to 310 μm in wet form, 100 to 310 μm in dry form |
£534.00
100g |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11544885
|
Cytiva
17-0061-01 |
1 mL/min. | 25 to 100 μm in wet form, 10 to 40 μm in dry form |
£669.00
100g |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
Description
- Available in two particle size ranges
Fine particle size
- Particle size, dry: 40 to 120μm
- Fractionation range, globular proteins: 4000 to 150,000
- Particle size, dry: 10 to 40μm
- Fractionation range, globular proteins: 4000 to 100,000
Spécification
100g | |
Sephadex G-100 Media | |
Useful for desalting and Buffer exchange of very large biomolecules | |
2 to 10 | |
Large biomolecules |
15 to 20mL/g in Distilled Water | |
Solid Bead | |
0.096 bar | |
Cross-linked Dextran | |
4° to 30°C |