missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G10 Media
Gel filtration medium for desalting and buffer exchange of peptides and small biomolecules
£259.00 - £1144.00
Specifications
Flow Rate | < Darcy´s Law |
---|---|
Particle Size | 55 to 165 μm in wet form, 40 to 120 μm in dry form |
Bed Volume | 2 to 3mL/g in Distilled Water |
Product Type | Sephadex G10 Media |
Format | Solid Bead |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11584865
|
Cytiva
17-0010-01 |
100g |
£259.00
100g |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
11594865
|
Cytiva
17-0010-02 |
500g |
£1144.00
500g |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
Description
- Beads behave as rigid spheres obeying Darcy's Law for purposes of determining maximum flowrate and maximum operating pressure
- Working pH range 2 to 13
- Particle size range 55 to 165μm
- Bed volume in distilled water 2-3mL/g
- Exclusionation limit: 7 x 102 dextrans, 7 x 102 globular proteins
Specifications
< Darcy´s Law | |
2 to 3mL/g in Distilled Water | |
Solid Bead | |
Desalting and Buffer exchange of Peptides and small biomolecules >700 M.W. | |
30cm | |
2 to 13 | |
Peptides and small biomolecules |
55 to 165 μm in wet form, 40 to 120 μm in dry form | |
Sephadex G10 Media | |
2.6cm | |
11.81 in. | |
24-49 cm/h, pressure drop cm H2O/bed height = 2, bed height 30 cm, 2.6 cm i.d | |
Cross-linked Dextran | |
4 to 30°C |