missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ Ion Exchange Media
Separate molecules over broad molecular weight and pH ranges
£365.00 - £1692.00
Specifications
For Use With (Equipment) | Column or Batch Techniques |
---|---|
Product Type | Sephadex Ion Exchange Media |
Format | Solid Spherical Bead |
For Use With (Application) | Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques |
Height (English) | 2 to 5.9 in. |
Product Code | Brand | Material | MWCO | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | MWCO | Quantity | Price | Quantity & Availability | |||
11504895
|
Cytiva
17-0200-01 |
QAE-Sephadex A-50 | 30,000 to 100,000 | 100g |
£365.00
100g |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
11584885
|
Cytiva
17-0180-02 |
DEAE-Sephadex A-50 | 30,000 to 100,000 | 500g |
£1682.00
500g |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11554885
|
Cytiva
17-0170-01 |
DEAE-Sephadex A-25 | Up to 30,000 | 100g |
£365.00
100g |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11564885
|
Cytiva
17-0170-02 |
DEAE-Sephadex A-25 | Up to 30,000 | 500g |
£1682.00
500g |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
11574885
|
Cytiva
17-0180-01 |
DEAE-Sephadex A-50 | 30,000 to 100,000 | 100g |
£370.00
100g |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11594885
|
Cytiva
17-0190-01 |
QAE-Sephadex A-25 | Up to 30,000 | 100g |
£365.00
100g |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11514895
|
Cytiva
17-0210-01 |
CM-Sephadex C-25 | Up to 30,000 | 100g |
£365.00
100g |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11524895
|
Cytiva
17-0210-02 |
CM-Sephadex C-25 | Up to 30,000 | 500g |
£1683.00
500g |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
11544895
|
Cytiva
17-0220-02 |
CM-Sephadex C-50 | 30,000 to 100,000 | 500g |
£1692.00
500g |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
11534895
|
Cytiva
17-0220-01 |
CM-Sephadex C-50 | 30,000 to 100,000 | 100g |
£368.00
100g |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
11554895
|
Cytiva
17-0230-01 |
SP-Sephadex C-25 | Up to 30,000 | 100g |
£368.00
100g |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
Description
Range of weak and strong anionic (A) and cationic (C) exchangers. Fractionation range: A-25 and C-25, up to 30 000; A-50 and C-50: 30 000–100 000.
Very high capacities even at elevated ionic strengths
Very low non-specific adsorption
Wide range of applications at laboratory and process scales
Suitable for column or batch techniques
Simple and economical to use
DEAE Sephadex
- Weak and strong anion exchanger
- pH range 2-9
QAE Sephadex
- Strong anion exchanger
- pH range: 2-12
- Weak cation exchanger
- pH range: 6-10
- Strong cation exchanger
- pH range: 4-13
Specifications
Column or Batch Techniques | |
Solid Spherical Bead | |
2 to 5.9 in. | |
Cross-linked Dextran |
Sephadex Ion Exchange Media | |
Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques | |
5 to 15cm | |
4° to 30°C |