missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ LH-20

Use for molecular sizing of natural product such as steroid, terpenoid, lipid, low molecular weight peptide. Cytiva Life Sciences™Sephadex™ LH-20 is a liquid chromatography media useful to analytical and industrial scale for preparation of closely related molecular species.

Brand:  Cytiva 17-0090-10

127.94 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 10309794

  • £142.00 / 25g
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepared for use in polar organic solvents and aqueous solvent mixtures
  • Made from hydroxypropylated dextran beads that have been crosslinked to yield a polysaccharide network
  • Hydrophilic and lipophilic dual nature of the matrix enables powerful chromatographic selectivity
  • Batch-to-batch reproducibility
  • Partical size dry: 18μ-111μm
  • Fractionation range globular proteins: 4x103 to 5 x 103

  • Stable in most aqueous and organic eluent systems
  • Shelf life: 5years
  • Autoclavable: 20 min at 121°C
  • Flow Velocity <∽12cm/min
  • pH stability working range: 2-13

Analytical and industrial scales for the preparation of closely related molecular species

TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 g
18 to 111 μm in dry form
Solid Spherical Bead
For fractionation of lipids, hormones, vitamins, and other small biomolecules in polar organic solvents and aqueous solvent mixtures
30cm
2 to 13
Steroid, Terpenoid, Lipid, low molecular weight Peptide
720 cm/hr.
Sephadex LH-20 Media
2.6cm
11.81 in.
25-45 cm/h, pressure drop cm H2O/bed height=2, beh height 30 cm, 2.6 cm i.d.
Hydroxypropylated, cross-linked Dextran
4 to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.