missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephaorose™ Fast Flow Anion Exchange Media: Q

Use for preparative protein separations in both research and industrial applications. Cytiva Q Sepharose™ Fast Flow Chromatography Media is a strong anion exchanger based on the well established Sepharose Fast Flow ion exchange platform.

Brand:  Cytiva 17-0510-10

76.00 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.



Product Code. 10383190

  • £82.40 / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
  • Matrix is composed of crosslinked 6% agarose beads
  • Part of Sepharose Fast Flow ion exchange platform
  • High chemical stability allows well proven cleaning-in-place (CIP) and sanitization protocols
  • Available in range of pre-packed process development tools

  • Ligand: quaternary amine
  • Ion exchanger type: strong anion exchanger
  • Ionic capacity: 0.18 to 0.25mmol Cl-/mL medium
  • Binding capacity: 120mg HSA/mL medium
  • Pressure/Flow specification: 400 to 700cm/hr., 100kPa, XK 50/30 column, bed height 15cm

Capture and purification of proteins.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mL
For Preparative Protein Separations in Both Research and Industrial AppliCatIons
Q Sepharose Fast Flow Media
45 to 165 μm
15 cm
Strong AnIon Exchanger
6% Cross-Linked Agarose with Dextran surface extenders
20% Ethanol
Stable in commonly used aqueous Buffers: 1 M NaOH, 8 M Urea, 8 M Guanidine Hydrochloride, 70% Ethanol
4°C to 30°C
400 to 700 cm/Hoμr
5.9 in.
0.18 to 0.24 mM Cl-/mL
100 kPa
2 to 12
Quaternary Amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    21

    Hours

    15

    Minutes

    34

    Seconds

    35
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

    Days

    52

    Hours

    15

    Minutes

    34

    Seconds

    35
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: Q > Q Sepharose; Pack size: 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.