Learn More
Ertugliflozin, MedChemExpress
MedChemExpress Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Brand: Medchem Express HY-15461
Description
Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.Specifications
Ertugliflozin | |
White | |
C22H25ClO7 | |
PF-04971729 | |
ClC1=CC=C([C@]23O[C@@](CO)(CO3)[C@@H](O)[C@H](O)[C@H]2O)C=C1CC4=CC=C(OCC)C=C4 | |
436.88 | |
Research | |
Metabolism-sugar/lipid metabolism |
1210344-57-2 | |
Research | |
5 mg | |
DMSO : 250 mg/mL (572.24 mM; Need ultrasonic) | |
436.88 | |
98.82% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed.