missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Septum for Magnetic Universal Precision Thread Cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand: Fisherbrand™ 17 02 1318
598.50 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
- 17mm Septa
Specifications
Septa | |
17 mm | |
1000 |
Blue, White | |
PTFE, Silicone | |
1.5 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction