missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophilic
Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
Brand: Cytiva 24152105050350
1293.72 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophilic bead based on crosslinking requirements for the encapsulation layer
Specifications
5% solids (50mg/mL) | |
DNA Sample Prep | |
1 μm |
2°C to 8°C | |
100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction