missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophilic
Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
£331.00 - £8076.92
Specifications
Concentration | 5% solids (50mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11588712
|
Cytiva
24152105050250 |
15 mL |
£331.00
Each |
In Stock
Log in to see stock. |
|||||
11834982
|
Cytiva
24152105050350 |
100 mL |
£1674.00
Each |
In Stock
Log in to see stock. |
|||||
11844982
|
Cytiva
24152105050450 |
1 L | N/A | ||||||
Description
- High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophilic bead based on crosslinking requirements for the encapsulation layer
Specifications
5% solids (50mg/mL) | |
2°C to 8°C | |
1 μm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title