missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag SpeedBeads™ Carboxyl Magnetic Beads, hydrophilic

Excellent for coupling oligos or proteins directly to the beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.

Brand:  Cytiva 45152105050250

409.91 GBP valid until 2024-12-31
Use promo code "21948" to get your promotional price.


 View more versions of this product

Product Code. 11548692

  • £459.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Double shell magnetite beads
  • . High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophilic bead based on crosslinking requirements for the encapsulation layer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5% solids (50mg/mL)
DNA Sample Prep, Protein Purification
1 μm
2°C to 8°C
15 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.