missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag SpeedBeads™ Carboxyl Magnetic Beads, hydrophilic
Excellent for coupling oligos or proteins directly to the beads. Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
£446.00 - £9230.77
Specifications
Concentration | 5% solids (50mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | DNA Sample Prep, Protein Purification |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11548692
|
Cytiva
45152105050250 |
15 mL |
£446.00
Each |
In Stock
Log in to see stock. |
|||||
11809792
|
Cytiva
45152105050350 |
100 mL |
£1853.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
13223609
|
Cytiva
45152105050450 |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Double shell magnetite beads
- . High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophilic bead based on crosslinking requirements for the encapsulation layer
Specifications
5% solids (50mg/mL) | |
2°C to 8°C | |
DNA Sample Prep, Protein Purification | |
1 μm |