Learn More
Cytiva Sera-Mag SpeedBeads™ Carboxyl Magnetic Beads, hydrophilic
Excellent for coupling oligos or proteins directly to the beads. Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
Brand: Cytiva 45152105050350
1711.25 GBP valid until 2024-12-31
Use promo code "21948" to get your promotional price.
Code : LE
Additional Details : Weight : 0.31750kg
Description
- Double shell magnetite beads
- . High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophilic bead based on crosslinking requirements for the encapsulation layer
Spécification
5% solids (50mg/mL) | |
DNA Sample Prep, Protein Purification | |
1 μm |
2°C to 8°C | |
100 mL |