missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SeraSil-Mag Silica Coated Superparamagnetic Beads
SeraSil-Mag beads for nucleic acid isolation deliver high purity DNA extraction in NGS applications.
£86.07 - £513.94
Specifications
Concentration | 20 mg/mL in water (0.05% sodium azide) |
---|---|
Content And Storage | Room Temperature |
Description | Storage Solution: Water + 0.05% sodium azide |
For Use With (Application) | Chaotropic salt chemistry and vortex mixing, roller mixing and sonication |
Product Type | Silica coated, core-shell structure |
Product Code | Brand | Quantity | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Size | Price | Quantity & Availability | ||||
16437655
|
Cytiva
29357369 |
5 mL | 400 nm diameter particles |
£106.56
5mL |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16447655
|
Cytiva
29357371 |
60 mL | 400 nm diameter particle |
£513.94
60mL |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16457655
|
Cytiva
29357373 |
5 mL | 700 nm diameter particle |
£86.07
5mL |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16467655
|
Cytiva
29357374 |
60 mL | 700 nm diameter particle |
£429.31
60mL |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Specifications
20 mg/mL in water (0.05% sodium azide) | |
Storage Solution: Water + 0.05% sodium azide | |
Silica coated, core-shell structure |
Room Temperature | |
Chaotropic salt chemistry and vortex mixing, roller mixing and sonication |