Learn More
BINDER™ Series CBF CO2 Incubator, with Hot Air Sterilization and Humidity Regulation



The CBF is the high-end CO2 incubator with humidity control that is suitable for all sensitive incubation applications in GMP environments. It ensures consistent results including in cell-based assays with multiwell plates.
Brand: BINDER™ 9640-0021
12652.19 GBP valid until 2025-12-31
Use promo code "25039" to get your promotional price.
View more versions of this product
Description
This BINDER incubator is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.- Contamination-safe humidification system without water pan
- Tried-and-tested anti-contamination concept with 180°C hot air sterilization
- Hot-air sterilizable CO2 sensor with single-beam infrared technology
- Seamless inner chamber made of stainless steel with flanges as shelf support system
- Fanless interior with Venturi CO2 gas mixing nozzle
- Intuitive touchscreen controller
- Internal data logger, measured values can be read out in open format via USB
- Troubleshooting system with visual and acoustic alarms
- Interfaces: Ethernet, USB, zero-voltage alarm contact
- Fail-safe CO2 system for protecting the pH of cell cultures
Specifications
9.4 cu. ft. | |
10 kg | |
267 L | |
0 to 20% | |
Fanless interior | |
None | |
Stainless Steel | |
3 | |
None | |
90 to 95% | |
CO2 Incubator | |
CBF260-230 V | |
IR sensor | |
745 x 885 x 1020 mm | |
230 V | |
7 A |
22 lb. | |
CBF260-230 V, Standard, Series CBF - CO2 incubators with hot air sterilization and humidity regulation, 267 L, 230 V 1 ∼ 50/60 Hz | |
Stainless Steel | |
IR Sensor | |
Natural | |
No Lighting | |
1 | |
1 to 21% | |
Nema 5-20P | |
Ambient +6° to 60°C | |
TFT touch | |
IR sensor | |
620 x 575 x 750 mm | |
125 kg | |
1.5 kW | |
50/60 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.