missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Serrated Pestles

Disperse homogenate into a mortar more efficiently with a serrated pestle. Kartell™ Serrated Pestles are ideal for tough materials such as tumors and tissue.

Brand:  Kartell™ 0630500

 View more versions of this product

Product Code. 10547862

  • £42.30 / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • PTFE head with Stainless Steel shaft
  • For use with borosilicate mortar for sample crushing
  • Ideal for tough materials such as tumors
Specifications

Specifications

5 mL
PTFE (head) Stainless Steel (shaft)
235 mm
Serrated Pestle
Tissue Grinder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ Serrated Pestles > Capacity (Metric): 5mL, Height (Metric): 235mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.