missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Serum Vial Racks

Designed to withstand elevated water temperatures for incubation.

Brand:  Bel-Art™ F18513-0072

41.33 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 15599991

  • £43.50 / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with serum vials (10-13 mm dia.), RIA vials and T3/T4 reaction vials (13-16 mm and 16-20 mm dia.)
  • Long-lasting and corrosion-resistant
  • With end plates handles
  • Made from polypropylene
  • Steam autoclavable at 121°C (250°F)
Specifications

Specifications

End plate handles
Corrosion resistant
White
Polypropylene
Rectangular
No
For use with serum vials
23.8 x 11.1 x 8.9cm
102 mm
13 mm
Vial Rack
Autoclavable
Tubes (10 to 13 mm)
72
2 Tiers
Non-sterile
L x W x H: 23.8 x 11.1 x 8.9 cm
241 mm
89 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Serum Vial Racks > Holds 10 - 13mm tubes; 72 places

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.