Learn More
Tocris Bioscience™ SET 2
Selective TRPV2 antagonist
Brand:  Tocris Bioscience™ 6990/10
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18034142
  • £164.89 / 10mg
Estimated Shipment: 27-08-2024
to see stock.
Description
Description
SET 2 is a TRPV2 (transient receptor potential vanilloid type 2) antagonist (IC50 = 0.46 μM). Selective for TRPV2 over TRPV1, TRPV3 and TRPV4 expressed in HEK293 cells. Inhibits lyosphosphatidic acid (LPA)-induced prostate cancer cell migration in vitro.
Specifications
Specifications
N-(2-Furanylmethyl)-3-[[4-(methylpropylamino)-6-(trifluoromethyl)-2-pyrimidinyl]thio]propanamide
10 mg
C17H21F3N4O2S
2313525-20-9
TRPV Antagonists
0.98