missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SET 2
Selective TRPV2 antagonist
£164.89 - £621.95
Specifications
Chemical Name or Material | N-(2-Furanylmethyl)-3-[[4-(methylpropylamino)-6-(trifluoromethyl)-2-pyrimidinyl]thio]propanamide |
---|---|
CAS | 2313525-20-9 |
Target | TRPV Antagonists |
Molecular Formula | C17H21F3N4O2S |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18065742
|
Tocris Bioscience™
6990/50 |
50 mg |
£621.95
50mg |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18034142
|
Tocris Bioscience™
6990/10 |
10 mg |
£164.89
10mg |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SET 2 is a TRPV2 (transient receptor potential vanilloid type 2) antagonist (IC50 = 0.46 μM). Selective for TRPV2 over TRPV1, TRPV3 and TRPV4 expressed in HEK293 cells. Inhibits lyosphosphatidic acid (LPA)-induced prostate cancer cell migration in vitro.Specifications
N-(2-Furanylmethyl)-3-[[4-(methylpropylamino)-6-(trifluoromethyl)-2-pyrimidinyl]thio]propanamide | |
TRPV Antagonists | |
0.98 |
2313525-20-9 | |
C17H21F3N4O2S |