Learn More
Tocris Bioscience™ SET 2
Selective TRPV2 antagonist
Brand: Tocris Bioscience™ 6990/50
Additional Details : Weight : 0.00970kg
Description
SET 2 is a TRPV2 (transient receptor potential vanilloid type 2) antagonist (IC50 = 0.46 μM). Selective for TRPV2 over TRPV1, TRPV3 and TRPV4 expressed in HEK293 cells. Inhibits lyosphosphatidic acid (LPA)-induced prostate cancer cell migration in vitro.Specifications
N-(2-Furanylmethyl)-3-[[4-(methylpropylamino)-6-(trifluoromethyl)-2-pyrimidinyl]thio]propanamide | |
50 mg | |
C17H21F3N4O2S |
2313525-20-9 | |
TRPV Antagonists | |
0.98 |