Learn More
Seward™ Stomacher™ Bag Racks
Seward has developed a full range of Stomacher™ Accessories to help provide a safe and efficient working environment in your laboratory.
Brand:  Seward™ BA6090
 View more versions of this product
Product Code. 11311434
  • £104.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Help to organize sample bags
  • High-grade plastic polymer construction
  • Provide perfect storage for Stomacher bag sample handling for a cleaner, more productive work environment
  • Perfect for loading closure bags pre-enrichment into the incubator
  • Easily dismantled and cleaned with most lab disinfectants (not autoclavable)
Specifications
Specifications
Bag Rack
Not Autoclavable
Bags
10
To prepare, store and transport samples in microbiology sample preparation
1
30cm x 11cm x 12cm
0.9kg
Sample bag rack (10 places)
Clear
Polymer
Non-sterile
Stomacher 80Biomaster
1
11 cm