Learn More
Cytiva SFX-Insect™ Cell Culture Media
Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.
Brand: Cytiva SH30278.LS
318.57 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : A0
Additional Details : Weight : 0.50000kg
Description
- Protein free media
Specifications
cGMP | |
Liquid | |
0.2 μm Sterile-filtered | |
No Protein, No Phenol Red, No HEPES |
2° to 8°C, away from light | |
Insect Cell Medium | |
Glutamine, Sodium Bicarbonate, Poloxamer 188 | |
6 x 1000 mL |