Learn More
Cytiva SFX-Insect™ Cell Culture Media
Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.
£66.90 - £386.00
Product Code | Brand | Quantity | Form | With Additives | Without Additives | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Form | With Additives | Without Additives | Price | Quantity & Availability | ||
10024393
|
Cytiva
SH30278.LS |
6 x 1000 mL | Liquid | Glutamine, Sodium Bicarbonate, Poloxamer 188 | No Protein, No Phenol Red, No HEPES |
£386.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10531813
|
Cytiva
SH30350.02 |
5 L | Powder | Glutamine, Poloxamer 188 | No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES |
£124.00
5 L |
Estimated Shipment: 25-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10673143
|
Cytiva
SH30278.02 |
1000 mL | Liquid | Glutamine, Poloxamer 188 | No Protein, No Phenol Red, No HEPES |
£66.90
1000mL |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11561911
|
Cytiva
SH30350.03 |
10 L | Powder | Glutamine, Poloxamer 188 | No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES |
£239.00
10 L |
Estimated Shipment: 25-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Protein-free media
Specifications
cGMP | |
Insect Cell Medium |