Learn More
Tocris Bioscience™ SGC 707
Potent and selective allosteric inhibitor of PRMT3
Brand: Tocris Bioscience™ 5367/50
373.21 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Description
SGC 707 is a potent allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 31 nM). Inhibits PRMT3-mediated methyltransferase activity in vitro. Displays >100-fold selectivity against 31 other methyltransferases and 250 other non-epigenetic targets. Decreases H4R3me2a levels in HEK293 cells in vitro.Negative control also available.
Specifications
>98% | |
SGC 707 | |
SGC 707 |
50 mg | |
298.34 |
For Research Use Only