Learn More
Tocris Bioscience™ SGC 707
Potent and selective allosteric inhibitor of PRMT3
£125.00 - £467.00
Specifications
Purity | >98% |
---|---|
Product Type | SGC 707 |
Molecular Weight (g/mol) | 298.34 |
Inhibitors | SGC 707 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762138
|
Tocris Bioscience™
5367/10 |
10 mg |
£125.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759917
|
Tocris Bioscience™
5367/50 |
50 mg |
£467.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SGC 707 is a potent allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 31 nM). Inhibits PRMT3-mediated methyltransferase activity in vitro. Displays >100-fold selectivity against 31 other methyltransferases and 250 other non-epigenetic targets. Decreases H4R3me2a levels in HEK293 cells in vitro.Negative control also available.
Specifications
>98% | |
SGC 707 | |
298.34 | |
SGC 707 |
For Research Use Only