Learn More
Trajan™ Methyl Deactivated Fused Silica Capillary Tubing
Essential for maintaining a high performance chromatography system.Trajan Scientific and Medical™ Methyl Deactivated Fused Silica Capillary Tubing is chemically inert and thermally stable up to 380°C.
£30.20 - £482.00
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Diameter (Metric) Inner | Overall Length (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Diameter (Metric) Inner | Overall Length (English) | Price | Quantity & Availability | ||
15228813
|
Trajan™
0624469 |
0.008 in. | 0.01 in. | 0.22 mm | 6.56 ft. |
£30.20
2m |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
15298803
|
Trajan™
0624461 |
0.005 in. | 0.008 in. | 0.15 mm | 16.4 ft. |
£64.00
5m |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15218813
|
Trajan™
0624465 |
0.005 in. | 0.014 in. | 0.15 mm | 6.56 ft. |
£30.20
2m |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15238803
|
Trajan™
0624450 |
0.002 in. | 0.007 in. | 0.075 mm | 6.56 ft. |
£30.20
2m |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15288803
|
Trajan™
0624460 |
0.005 in. | 0.008 in. | 0.15 mm | 6.56 ft. |
£30.20
2m |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15238813
|
Trajan™
0624470 |
0.012 in. | 0.016 in. | 0.32 mm | 6.56 ft. |
£30.20
2m |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
12955751
|
Trajan™
0624471 |
0.012 in. | 0.016 in. | 0.32 mm | 16.4 ft. |
£66.95
5m |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
15288813
|
Trajan™
0624479 |
0.020 in. | 0.026 in. | 0.53 mm | 6.56 ft. |
£30.20
2m |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15298813
|
Trajan™
062449 |
0.006 in. | 0.01 in. | 0.17 mm | 16.4 ft. |
£64.00
5m |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
15248813
|
Trajan™
0624473 |
0.012 in. | 0.016 in. | 0.32 mm | 82 ft. |
£281.00
25m |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
10340661
|
Trajan™
064050 |
0.008 in. | 0.01 in. | 0.22 mm | 16.4 ft. |
£241.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
10218580
|
Trajan™
064054 |
0.020 in. | 0.026 in. | 0.53 mm | 16.4 ft. |
£446.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
10781802
|
Trajan™
064052 |
0.012 in. | 0.016 in. | 0.32 mm | 16.4 ft. |
£241.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
12945751
|
Trajan™
062447 |
0.008 in. | 0.01 in. | 0.22 mm | 16.4 ft. |
£74.40
5m |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
15258853
|
Trajan™
064034 |
0.020 in. | 0.026 in. | 0.53 mm | 82 ft. |
£482.00
25m |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
11997754
|
Trajan™
0624431 |
0.25 mm | 0.01 in. | 0.001 in. | 6.6 ft. |
£30.20
Each |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||
15248853
|
Trajan™
064033 |
0.53 mm | 0.03 in. | 0.02 in. | 32.8 ft. |
£278.00
10m |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||
Description
- Suitable for organic and aqueous solvents
- Methyl deactivated for use with hydrocarbons (pentane, hexane, heptane, iso-octane, aromatics and mixed solvents - n-paraffins/chlorinated solvents
- Individually tested performance of each 30m length from which all shorter lengths are prepared
- Ideal for biotechnology applications
Specifications
Capillary Tubing |